Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butyl isocyanide

Catalog No. 1332801G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-328-01G 1 g
13-328-05G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-328-01G Supplier Sigma Aldrich Supplier No. 1332801G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Butyl isocyanide was used to study the crystal structure of carbon monoxide dehydrogenases-ii isolated from carboxydothermus hydrogenoformans
  • It was used to probe the mechanism of no activation of the hemoprotein soluble guanylate cyclase
  • .butyl isocyanide forms complex with ferric and ferrous iron of the heme in cytochrome p450.may form haziness and/or precipitate with no loss in purity

Specifications

CAS 2769-64-4
Density 0.795 g/mL (at 25°C (literature))
Molecular Formula C5H9N
Refractive Index n20/D 1.396 (literature)
Linear Formula CH3(CH2)3 NC
MDL Number MFCD00000008
Quantity 1 g
Molecular Weight (g/mol) 83.13
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.