Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Isothiocyanate Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022425 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2425 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2425 Supplier MilliporeSigma Supelco Supplier No. 16313250MG

missing translation for 'validMainframeMsg'

Specifications

CAS 592-82-5
Density 0.955 g/mL (at 25°C (literature))
Boiling Point 70°C to 71°C (35 mmHg) (literature)
Flash Point 66°C (Closed Cup)
Molecular Formula C5H9NS
Linear Formula CH3(CH2)3 NCS
Quantity 250 mg
MDL Number MFCD00004824
UN Number UN 3264 8 / PGII
Synonym NSC 194808
Formula Weight 115.20
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.