Learn More
Medchemexpress LLC Butyl Laurate 100g
SDP
Catalog No. 5000206684
Encompass_Preferred
Butyl Laurate is an ester compound with a fruity odor commonly used as a flavoring agent in the food and beverage industry In addition it is used as a solvent plasticizer or lubricant in various industrial applications Its unique chemical properties make it an important ingredient in a variety of commercial products including cosmetics personal care products and cleaners
Catalog No. 50-002-06684 Supplier Medchemexpress LLC Supplier No. HYW127330100G
Butyl Laurate is an ester compound with a fruity odor commonly used as a flavoring agent in the food and beverage industry In addition it is used as a solvent plasticizer or lubricant in various industrial applications Its unique chemical properties make it an important ingredient in a variety of commercial products including cosmetics personal care products and cleaners