Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI AMERICA Butyl Methacrylate, Reagent, >99.0% (GC), 500 mL

Catalog No. 502780937
Encompass_Preferred

Butyl Methacrylate is a reagent-grade monomer utilized in polymer synthesis and pharmaceutical research. This compound is provided as a stabilized, colorless clear liquid.

  • Purity of >99.0% by gas chromatography.
  • Chemical formula is C8H14O2 with a molecular weight of 142.20.
  • Supplied in a 500 mL clear glass bottle.
  • Recommended storage in a cool and dark place, preferably below 15°C.
  • Hazardous material requiring specific shipping considerations (UN2227).

Catalog No. 50-278-0937 Supplier Grainger Supplier No. 19KR97
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Butyl Methacrylate is a reagent-grade monomer utilized in polymer synthesis and pharmaceutical research. This compound is provided as a stabilized, colorless clear liquid.

  • Purity of >99.0% by gas chromatography.
  • Chemical formula is C8H14O2 with a molecular weight of 142.20.
  • Supplied in a 500 mL clear glass bottle.
  • Recommended storage in a cool and dark place, preferably below 15°C.
  • Hazardous material requiring specific shipping considerations (UN2227).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.