Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl N-(3-Trifluoromethylphenyl)anthranilate 98.0+%, TCI America™
SDP

Catalog No. B5391200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5391200MG 200 mg
B53911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5391200MG Supplier TCI America Supplier No. B5391200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67330-25-0
Molecular Formula C18H18F3NO2
Molecular Weight (g/mol) 337.342
MDL Number MFCD00866001
InChI Key JDLSRXWHEBFHNC-UHFFFAOYSA-N
Synonym N-(3-Trifluoromethylphenyl)anthranilic Acid Butyl Ester, Butyl Flufenamate, Flufenamic Acid Butyl Ester, Butyl 2-[[3-(Trifluoromethyl)phenyl]amino]benzoate, 2-[[3-(Trifluoromethyl)phenyl]amino]benzoic Acid Butyl Ester, Ufenamate
PubChem CID 5632
IUPAC Name butyl 2-[3-(trifluoromethyl)anilino]benzoate
SMILES CCCCOC(=O)C1=CC=CC=C1NC2=CC=CC(=C2)C(F)(F)F

Specifications

Color Yellow
Boiling Point 170°C
Quantity 200 mg
Formula Weight 337.34
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material Butyl N-(3-Trifluoromethylphenyl)anthranilate
RTECSNumber DH8511000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.