Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butyl nitrite

Catalog No. 226637100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-663-7100G 100 g
22-663-725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-663-7100G Supplier Sigma Aldrich Supplier No. 226637100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The photodissociation dynamics of butyl nitrite was studied using time-resolved fourier transform infrared (tr-ftir) emission spectroscopy
  • The effects of butyl nitrite on methyl cobalamin and 5-methyl tetrahydrofolate were studied

Specifications

CAS 544-16-1
Density 0.882 g/mL (at 25°C (literature))
Boiling Point 78°C (lit.)
Molecular Formula C4H9NO2
Refractive Index n20/D 1.376 (literature)
Linear Formula CH3(CH2)3 ONO
MDL Number MFCD00002058
Quantity 100 g
Molecular Weight (g/mol) 103.12
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.