Learn More
Medchemexpress LLC Butyl Palmitate 50g
SDP
Catalog No. 5000206502
Encompass_Preferred
Butyl Palmitate is an ester compound commonly used as a conditioning emollient or fragrance in a variety of cosmetic and personal care products In addition it can be used as a solvent or lubricant in various industrial applications Its unique chemical properties make it an important ingredient in a variety of commercial products including perfumes lotions and hair care products
Catalog No. 50-002-06502 Supplier Medchemexpress LLC Supplier No. HYW12733850G
Butyl Palmitate is an ester compound commonly used as a conditioning emollient or fragrance in a variety of cosmetic and personal care products In addition it can be used as a solvent or lubricant in various industrial applications Its unique chemical properties make it an important ingredient in a variety of commercial products including perfumes lotions and hair care products