Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butyl phenyl ether

Catalog No. 142344100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-234-4100G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-234-4100G Supplier Sigma Aldrich Supplier No. 142344100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Butyl phenyl ether

Specifications

CAS 1126-79-0
Melting Point -19°C (lit.)
Density 0.935 g/mL (at 25°C (literature))
Boiling Point 210.3°C (lit.)
Molecular Formula C10H14O
Refractive Index n20/D 1.497 (literature)
Linear Formula C6H5O(CH2)3 CH3
MDL Number MFCD00009438
Quantity 250 mg
Molecular Weight (g/mol) 150.22
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.