Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Phthalyl Butyl Glycolate 95.0+%, TCI America™
SDP

Catalog No. B073725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B073725G 25 g
B0737500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B073725G Supplier TCI America Supplier No. B073725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85-70-1
Molecular Formula C18H24O6
Molecular Weight (g/mol) 336.384
MDL Number MFCD00053793
InChI Key GOJCZVPJCKEBQV-UHFFFAOYSA-N
Synonym butyl phthalyl butyl glycolate, butoxycarbonylmethyl butyl phthalate, reomol 4pg, butyl carbobutoxymethyl phthalate, santicizer b-16, 2-butoxy-2-oxoethyl butyl phthalate, butyl glycolyl butyl phthalate, butyl phthalate butyl glycolate, caswell no. 131b, dibutyl o-carboxybenzoyloxyacetate
PubChem CID 6819
IUPAC Name 2-O-(2-butoxy-2-oxoethyl) 1-O-butyl benzene-1,2-dicarboxylate
SMILES CCCCOC(=O)COC(=O)C1=CC=CC=C1C(=O)OCCCC

Specifications

Color Yellow
Boiling Point 345°C
Quantity 25 g
Formula Weight 336.38
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Phthalyl Butyl Glycolate
EINECSNumber (3)-1314
RTECSNumber TI0535000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.