Learn More
AdipoGen Butyl-pyridinio-propanesulfona
SDP
Supplier:  AdipoGen CDXB0109G003
Encompass_Preferred
Chemical. CAS 570412-84-9. Formula C12H19NO3S. MW 257.35. Synthetic. NDSB-256-4T is a non-detergent sulfobetaine NDSB that prevents protein aggregation and facilitates protein-folding, presumably by interacting with the early folding intermediates. Shown to improve the in vitro renaturation efficiency of reduced hen lysozyme and chemically unfolded tryptophan synthase beta2 subunit.
Catalog No. 50-256-1676