Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Butyl Rubber Hood (Formerly 522-02-23R01)

Catalog No. 19071192 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-071-192 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-071-192 Supplier 3M™ Supplier No. 7000131727
  • $1,458.00
    $1,139.65 / Case of 1

    Save $318.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used with 3M™ Breathe Easy™ Powered Air Purifying Respirator (PAPR) Systems

  • Used for First Receiver, patient decontamination programs
  • Approved to the chemical, biological, radiological, and nuclear (CBRN) loose-fitting PAPR standard developed by the National Institute for Occupational Safety andHealth (NIOSH) for gases and vapors
  • The high efficiency filters in a CBRN cartridge filter out 99.97% of particulates
  • 3M Catalog# BE-10BR

TRUSTED_SUSTAINABILITY

Specifications

Assigned Protection Factor (APF) 1,000
Color Green
Material Butyl Rubber
Product Type Headgear
Size Standard
For Use With (Application) Defense, Military, Law Enforcement, Emergency Preparedness
For Use With (Equipment) 3M™ Breathe Easy™ Powered Air Purifying Respirator (PAPR) Systems
Model BE-10BR
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.