Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp™ Scientific Butyl Seal for SER158
SDP

Catalog No. 11392085
Change view
Click to view available options
For Use With (Equipment):
SER158/3 Solvent Extractor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-392-085 SER158/3 Solvent Extractor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-392-085 Supplier Velp™ Scientific Supplier No. A00000298
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Butyl seal for use with the SER158 Solvent Extractor.

  • Prevent leaks with these butyl seals

Specifications

Product Type Butyl Seal
For Use With (Equipment) SER158/3 Solvent Extractor
Color Gray
Quantity 3 Seals
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.