Learn More
Sigma Aldrich Fine Chemicals Biosciences Butyl SepharoseR High Perf
Catalog No. 502907278
Encompass
Butyl Sepharose High Performance is an aliphatic hydrophobic interaction chromatography (HIC) medium designed for intermediate and polishing step purification steps when high reSOLUTIONon has priority
Catalog No. 50-290-7278 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17543202
Butyl Sepharose High Performance is an aliphatic hydrophobic interaction chromatography (HIC) medium designed for intermediate and polishing step purification steps when high reSOLUTIONon has priority