Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butyl stearate

Catalog No. 857201L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-720-1L 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 85-720-1L Supplier Sigma Aldrich Supplier No. 857201L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Butyl stearate

Specifications

CAS 123-95-5
Melting Point 17°C to 22°C
Density 0.861 g/mL (at 20°C (literature))
Molecular Formula C22H44O2
Refractive Index n20/D 1.443
Linear Formula CH3(CH2)16 COO(CH2)3 CH3
MDL Number MFCD00026669
Quantity 100 mg
Synonym Stearic acid butyl ester
Molecular Weight (g/mol) 340.58
Percent Purity 40-60% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.