Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Valerate 98.0+%, TCI America™
SDP

Catalog No. B432225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B432225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B432225ML Supplier TCI America Supplier No. B432225ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 591-68-4
Color Yellow
Boiling Point 185°C
Molecular Formula C9H18O2
Quantity 25 mL
MDL Number MFCD00015258
Synonym Valeric Acid Butyl Ester, Butyl Pentanoate, Pentanoic Acid Butyl Ester
InChI Key OKJADYKTJJGKDX-UHFFFAOYSA-N
SMILES CCCCC(=O)OCCCC
IUPAC Name butyl pentanoate
Molecular Weight (g/mol) 158.241
PubChem CID 61137
Formula Weight 158.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Valerate
Show More Show Less
EINECSNumber (2)-0776
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.