Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butylamine

Catalog No. 47130525L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-130-525L 2.5 L
47-130-51L 1 L
47-130-5250ML 250 mL
47-130-525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 47-130-525L Supplier Sigma Aldrich Supplier No. 4713052.5L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Butylamine

Specifications

CAS 109-73-9
Melting Point -49°C
Density 0.74 g/mL at 25°C
Boiling Point 78°C
Molecular Formula C4H11N
Refractive Index n20/D 1.401
Linear Formula CH3(CH2)3NH2
MDL Number MFCD00011690
Quantity 2.5 L
Synonym 1-Aminobutane; n-Butylamine
Molecular Weight (g/mol) 73.14 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.