Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butylamine Hydrobromide 98.0+%, TCI America™
SDP

Catalog No. B51865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51865G 5 g
B51861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51865G Supplier TCI America Supplier No. B51865G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15567-09-6
Molecular Formula C4H12BrN
Molecular Weight (g/mol) 154.051
InChI Key SXGBREZGMJVYRL-UHFFFAOYSA-N
Synonym Butylammonium Bromide
PubChem CID 12199058
IUPAC Name butan-1-amine;hydrobromide
SMILES CCCCN.Br

Specifications

Color White
Quantity 5 g
Formula Weight 154.05
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Butylamine Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.