Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butylated hydroxyanisole

Catalog No. 501754728
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-175-4728 5 g
50-175-4726 100 g
50-175-4727 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-175-4728 Supplier Sigma Aldrich Supplier No. B12535G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Butylated hydroxyanisole
TRUSTED_SUSTAINABILITY

Specifications

CAS 25013-16-5
Melting Point 58°C to 60°C (lit.)
Molecular Formula C11H16O2
Linear Formula (CH3)3 CC6H3(OCH3)OH
Quantity 5 g
MDL Number MFCD01779059
Synonym 2(3)-t-Butyl-4-hydroxyanisole; 2(3)-t-Butylhydroquinone monomethyl ether; BHA
Molecular Weight (g/mol) 180.24
Percent Purity ≥98.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.