Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butylated hydroxyanisole, ≥99.0% (sum of isomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111013696 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3696 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3696 Supplier MilliporeSigma Supelco Supplier No. 78943500MG
Add to Cart
Add to Cart

Butylated hydroxyanisole is a lipophilic compound(3), which is is widely used as an antioxidant in food industry.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 599°F
CAS 25013-16-5
Vapor Density 6.2 (vs. air)
Flash Point 113°C - Closed Cup
Physical Form Neat
Linear Formula (CH3)3 CC6H3(OCH3)OH
MDL Number MFCD01779059
Melting Point 58°C to 60°C
Synonym Butylated hydroxyanisole; E320; 2(3)-t-Butyl-4-hydroxyanisole; 2(3)-t-Butylhydroquinone monomethyl ether; BHA
Type Analytical Standard
Quantity 500 mg
Formula Weight 180.24
Percent Purity ≥99.0% (sum of isomers, GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.