Learn More
Medchemexpress LLC HY-Y0172 100mg Medchemexpress, Butylated hydroxytoluene CAS:128-37-0 Purity:>98%
Supplier: Medchemexpress LLC HYY0172100MG
Medchemexpress, HY-Y0172 100mg Butylated hydroxytoluene CAS:128-37-0 Butylated hydroxytoluene is an antioxidant widely used in foods and in food-related products. Butylated hydroxytoluene is a Ferroptosis inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.