Learn More
Sigma Aldrich Fine Chemicals Biosciences Butylated hydroxytoluene, 128-37-0, MFCD00011644, 1 KG
Catalog No. NC1166226
Encompass
Linear Formula: ((CH3)3C)2C6H2(CH3)OH, Molecular Weight: 220.35, ≥99%, FCC, FG, Synonym: 2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC. Butylated hydroxytoluene is a synthetic phenolic compound mainly used as an antioxidant and preservative in the food industry. It is used to prevent the lipid oxidation in oils and fat-containing foods.
Catalog No. NC1166226 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2184051KGK
Linear Formula: ((CH3)3C)2C6H2(CH3)OH, Molecular Weight: 220.35, ≥99%, FCC, FG, Synonym: 2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC. Butylated hydroxytoluene is a synthetic phenolic compound mainly used as an antioxidant and preservative in the food industry. It is used to prevent the lipid oxidation in oils and fat-containing foods.