Learn More
Medchemexpress LLC Butylated hydroxytoluene-d21 | 64502-99-4 | MFCD00209760 | 98.7% | 241.48 | C15H3D21O | 5mg
SDP
Catalog No. 5000270170
Encompass_Preferred
Butylated hydroxytoluene-d21 is the deuterium labeled Butylated hydroxytoluene[1] Butylated hydroxytoluene is an antioxidant widely used in foods and in food-related products[2] Butylated hydroxytoluene is a Ferroptosis inhibitor[3]
Catalog No. 50-002-70170 Supplier Medchemexpress LLC Supplier No. HYY0172S5MG
Butylated hydroxytoluene-d21 is the deuterium labeled Butylated hydroxytoluene[1] Butylated hydroxytoluene is an antioxidant widely used in foods and in food-related products[2] Butylated hydroxytoluene is a Ferroptosis inhibitor[3]