Learn More
Medchemexpress LLC Butylboronic acid | 4426-47-5 | 101.94 | 1 G
SDP
Catalog No. 5000449737
Encompass_Preferred
Butylboronic acid is a competitive soybean urease inhibitor with an IC50 of 2.9 mM and a Ki of 1.5 mM. It is for research use only and promotes cell proliferation, alkaline phosphatase activity, and expression of GJA1, ALP, and NaBC1 in human dental pulp cells in vitro at 0.1 mM concentration (3-28 days).
  • Competitive soybean urease inhibitor
  • IC50 of 2.9 mM for soybean (Glycine max) urease
  • Ki of 1.5 mM for soybean (Glycine max) urease
  • Promotes cell proliferation, alkaline phosphatase activity, and expression of GJA1, ALP, and NaBC1 in human dental pulp cells in vitro at 0.1 mM concentration (3-28 days)
Catalog No. 50-004-49737 Supplier Medchemexpress LLC Supplier No. HYW0009321G
Butylboronic acid is a competitive soybean urease inhibitor with an IC50 of 2.9 mM and a Ki of 1.5 mM. It is for research use only and promotes cell proliferation, alkaline phosphatase activity, and expression of GJA1, ALP, and NaBC1 in human dental pulp cells in vitro at 0.1 mM concentration (3-28 days).
  • Competitive soybean urease inhibitor
  • IC50 of 2.9 mM for soybean (Glycine max) urease
  • Ki of 1.5 mM for soybean (Glycine max) urease
  • Promotes cell proliferation, alkaline phosphatase activity, and expression of GJA1, ALP, and NaBC1 in human dental pulp cells in vitro at 0.1 mM concentration (3-28 days)