Learn More
Medchemexpress LLC Butylboronic acid | 4426-47-5 | 101.94 | 10 MM * 1 ML
SDP
Catalog No. 5000449172
Encompass_Preferred
Butylboronic acid is a competitive soybean urease inhibitor with an IC50 of 2.9 mM and a Ki of 1.5 mM.
  • Competitive soybean urease inhibitor (IC50 of 2.9 mM and a Ki of 1.5 mM).
  • Promotes cell proliferation, alkaline phosphatase activity, and expression of GJA1, ALP, and NaBC1 in human dental pulp cells in vitro.
Catalog No. 50-004-49172 Supplier Medchemexpress LLC Supplier No. HYW00093210MM/1ML
Butylboronic acid is a competitive soybean urease inhibitor with an IC50 of 2.9 mM and a Ki of 1.5 mM.
  • Competitive soybean urease inhibitor (IC50 of 2.9 mM and a Ki of 1.5 mM).
  • Promotes cell proliferation, alkaline phosphatase activity, and expression of GJA1, ALP, and NaBC1 in human dental pulp cells in vitro.