Learn More
Medchemexpress LLC Butylboronic acid | 4426-47-5 | 101.94 | 5 G
SDP
Catalog No. 5000449722
Encompass_Preferred
Butylboronic acid is a competitive soybean urease inhibitor with an IC50 of 2.9 mM and a Ki of 1.5 mM. It exerts its inhibitory effect on soybean (Glycine max) urease through a competitive inhibition mechanism. Additionally, it promotes cell proliferation, alkaline phosphatase activity, and expression of GJA1, ALP, and NaBC1 in human dental pulp cells in vitro. This product is intended for research use only.
  • Competitive soybean urease inhibitor
  • IC50 of 2.9 mM (soybean (Glycine max) urease)
  • Ki of 1.5 mM
  • Promotes cell proliferation in human dental pulp cells
  • Enhances alkaline phosphatase activity
  • Increases expression of GJA1, ALP, and NaBC1
Catalog No. 50-004-49722 Supplier Medchemexpress LLC Supplier No. HYW0009325G
Butylboronic acid is a competitive soybean urease inhibitor with an IC50 of 2.9 mM and a Ki of 1.5 mM. It exerts its inhibitory effect on soybean (Glycine max) urease through a competitive inhibition mechanism. Additionally, it promotes cell proliferation, alkaline phosphatase activity, and expression of GJA1, ALP, and NaBC1 in human dental pulp cells in vitro. This product is intended for research use only.
  • Competitive soybean urease inhibitor
  • IC50 of 2.9 mM (soybean (Glycine max) urease)
  • Ki of 1.5 mM
  • Promotes cell proliferation in human dental pulp cells
  • Enhances alkaline phosphatase activity
  • Increases expression of GJA1, ALP, and NaBC1