Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butylboronic Acid (contains varying amounts of Anhydride) [for Esterification], TCI America™
SDP

Catalog No. B05295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0529-5G 5 g
B0529-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0529-5G Supplier TCI America Supplier No. B05295G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4426-47-5
Molecular Formula C4H11BO2
Molecular Weight (g/mol) 101.94
MDL Number MFCD00002106
InChI Key QPKFVRWIISEVCW-UHFFFAOYSA-N
Synonym n-butylboronic acid, 1-butylboronic acid, 1-butaneboronic acid, n-butaneboronic acid, boronic acid, butyl
PubChem CID 20479
IUPAC Name butylboronic acid
SMILES B(CCCC)(O)O

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 5 g
Formula Weight 101.94
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.