Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butylboronic Acid N,N,N',N'-Tetramethyl-D-Tartaric Acid Diamide Ester

Catalog No. 5948065G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-480-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 59-480-65G Supplier Sigma Aldrich Supplier No. 5948065G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Butylboronic acid N,N,N'sa,N'sa-tetramethyl-D-tartaric acid diamide is a dioxaborolane ligand that can be used to prepare:Enantioenriched spiropentanes via zinc catalyzed cyclopropanation of corresponding hydroxymethylallenes. 1,2,3-trisubstituted cyclopropane derivatives by asymmetric cyclopropanation of allylic alcohols in the presence of zinc reagents. Glycolipids plakosides A, B, and their derivatives by Sharpless asymmetric dihydroxylation and cyclopropanation reaction.

Specifications

CAS 258865-48-4
Melting Point 158°C to 162°C
Molecular Formula C29H44B2O4
Refractive Index n20/D 1.4780
Linear Formula (HO)2BC(C6H3C(C8H17)2C6H3B(OH)2
MDL Number MFCD03701608
Quantity 5 g
Synonym (4S,5S)-2-Butyl-N,N,N'N'-tetramethyl-1,3,2-dioxaborolane-4,5-dicarboxamide
Molecular Weight (g/mol) 478.28 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.