Learn More
Selleck Chemical LLC Butylhydroxyanisole-250MG
SDP
Catalog No. 503137759
Encompass_Preferred
Butylhydroxyanisole (Butylated hydroxyanisole BHA E320) is an inhibitor of ferroptosis a food preservative and antioxidant that prevents the formation of harmful compounds extending shelf life However it s associated with liver toxicity reproductive organ development issues learning delays and sleep problems
Catalog No. 50-313-7759 Supplier Selleck Chemical LLC Supplier No. E1925250MG
Butylhydroxyanisole (Butylated hydroxyanisole BHA E320) is an inhibitor of ferroptosis a food preservative and antioxidant that prevents the formation of harmful compounds extending shelf life However it s associated with liver toxicity reproductive organ development issues learning delays and sleep problems