Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butylone-d3 hydrochloride solution, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant

Catalog No. B0461ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0461ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0461ML Supplier MilliporeSigma Supplier No. B0461ML
Only null left
Add to Cart
Add to Cart

Also known as bk-MBDB or B1, butylone is a new designer drug with significant abuse reported in the US and Europe.

Also known as bk-MBDB or B1, butylone is a new designer drug with significant abuse reported in the US and Europe. This new internal standard is suitable for numerous GC/MS and LC/MS applications from clinical toxicology testing and forensic analysis to urine drug testing and isotope dilution methods.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butylone-D3 Hydrochloride
Packaging Ampule of 1 mL
CAS 1231710-63-6
Molecular Formula C12D3H12NO3 HCl
Concentration 100 μg/mL in Methanol (as free base)
For Use With (Equipment) Gas Chromatography (GC), Liquid Chromatography (LC)
Solution Type Standard Solution
Type Certified Reference Material
Industry Type Forensics, Toxicology
Molecular Weight (g/mol) 260.73
Quantity 1 mL
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.