Learn More
U.S. Pharmacopeia Butylparaben, 94-26-8, MFCD00016478, 200mg
Catalog No. NC9981237
Encompass
Molecular formula C11H14O3, Molecular weight 194.23, Melting Point 154.4 - 161.6 °F (68 - 72 °C), Boiling Point 312.8 - 314.6 °F (156 - 157 °C). Synonyms: Butyl parahydroxybenzoate, Butyl 4-hydroxybenzoate
Catalog No. NC9981237 Supplier U.S. Pharmacopeia Supplier No. 1084000
Molecular formula C11H14O3, Molecular weight 194.23, Melting Point 154.4 - 161.6 °F (68 - 72 °C), Boiling Point 312.8 - 314.6 °F (156 - 157 °C). Synonyms: Butyl parahydroxybenzoate, Butyl 4-hydroxybenzoate