Learn More
Medchemexpress LLC Butylparaben-d4 | 1219798-67-0 | 98.6% | 5 MG
SDP
Catalog No. 5000413675
Encompass_Preferred
Butylparaben-d4 is the deuterium labeled Butylparaben, an organic compound proven to be a successful antimicrobial preservative in cosmetics. It is also used in medication suspensions and as a flavoring additive in food. For research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-13675 Supplier Medchemexpress LLC Supplier No. HYB1431S5MG
Butylparaben-d4 is the deuterium labeled Butylparaben, an organic compound proven to be a successful antimicrobial preservative in cosmetics. It is also used in medication suspensions and as a flavoring additive in food. For research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS