Learn More
Medchemexpress LLC Butylparaben (standard) | 94-26-8 | 98.9% | 100 MG
SDP
Catalog No. 5000412913
Encompass_Preferred
Butylparaben (Standard) is the analytical standard of Butylparaben. It is an organic compound used as an antimicrobial preservative in cosmetics, in active molecule suspensions, and as a food flavoring additive. This product is intended for research and analytical applications, particularly as a reference standard in qualitative, quantitative, and methodological research experiments using techniques like HPLC, GC, and MS.
  • Analytical standard
  • Used as an antimicrobial preservative in cosmetics
  • Found in active molecule suspensions
  • Used as a food flavoring additive
  • Intended for research and analytical applications
  • Reference standard for qualitative, quantitative, and methodological research
  • Suitable for techniques such as HPLC, GC, and MS
Catalog No. 50-004-12913 Supplier Medchemexpress LLC Supplier No. HYB1431R100MG
Butylparaben (Standard) is the analytical standard of Butylparaben. It is an organic compound used as an antimicrobial preservative in cosmetics, in active molecule suspensions, and as a food flavoring additive. This product is intended for research and analytical applications, particularly as a reference standard in qualitative, quantitative, and methodological research experiments using techniques like HPLC, GC, and MS.
  • Analytical standard
  • Used as an antimicrobial preservative in cosmetics
  • Found in active molecule suspensions
  • Used as a food flavoring additive
  • Intended for research and analytical applications
  • Reference standard for qualitative, quantitative, and methodological research
  • Suitable for techniques such as HPLC, GC, and MS