Learn More
Selleck Chemical LLC Butylscopolamine Bromide S2378-25mg
SDP
Catalog No. 503112815
Encompass_Preferred
Butylscopolamine Bromide (Scopolamine Butylbromide (-)-Scopolamine butylbromide Hyoscine butylbromide) is a peripherally acting antimuscarinic anticholinergic agent used as an abdominal-specific antispasmodic
Catalog No. 50-311-2815 Supplier Selleck Chemical LLC Supplier No. S237825MG
Butylscopolamine Bromide (Scopolamine Butylbromide (-)-Scopolamine butylbromide Hyoscine butylbromide) is a peripherally acting antimuscarinic anticholinergic agent used as an abdominal-specific antispasmodic