Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butyltin Trichloride

Catalog No. 201057500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-105-7500G 500 g
20-105-75G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 20-105-7500G Supplier Sigma Aldrich Supplier No. 201057500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Butyltin Trichloride

Specifications

CAS 1118-46-3
Density 1.693 g/mL at 25°C
Boiling Point 93°C (10 mmHg, lit.)
Molecular Formula C4H9Cl3Sn
Refractive Index n20/D 1.523
Linear Formula CH3(CH2)3SnCl3
MDL Number MFCD00000515
Quantity 500 g
Synonym Butyltrichlorotin
Molecular Weight (g/mol) 282.18 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.