Learn More
Medchemexpress LLC Butyne-DOTA | 2125661-62-1 | MFCD35160514 | 97.0% | 455.51 | C20H33N5O7 | 10mg
SDP
Catalog No. 5000269479
Encompass_Preferred
Butyne-DOTAis a bifunctional chelator (Bifunctional Chelator BFC) and a macrocyclic DOTA derivative used for tumor pre-targeting Butyne-DOTA can be used for conjugation of peptides and radionuclides
Catalog No. 50-002-69479 Supplier Medchemexpress LLC Supplier No. HYW78207810MG
Butyne-DOTAis a bifunctional chelator (Bifunctional Chelator BFC) and a macrocyclic DOTA derivative used for tumor pre-targeting Butyne-DOTA can be used for conjugation of peptides and radionuclides