Learn More
Medchemexpress LLC Butyne-DOTA-tris(t-butyl ester) | 2125661-54-1 | 98.5% | 623.82 | C32H57N5O7 | 50 MG
SDP
Catalog No. 5000281743
Encompass_Preferred
Butyne-DOTA-tris(t-butyl ester) is a bifunctional chelator and macrocyclic DOTA derivative used for bioconjugation and tumor pre-targeting research. It contains tert-butyl protected carboxylate arms and a terminal butyne handle that enables click-style conjugation to peptides, proteins, or targeting vectors, facilitating subsequent radiolabeling or imaging workflows. The material is supplied as a white to off-white solid with high reported purity and recommended cold storage for long-term stability.
  • Bifunctional chelator suitable for bioconjugation
  • Terminal alkyne handle enables click chemistry conjugation
  • Tert-butyl protected carboxylates improve stability during synthesis
  • High purity suitable for radiolabeling workflows
  • Available in multiple pack sizes for research needs
  • Recommended cold storage to preserve integrity
Catalog No. 50-002-81743 Supplier Medchemexpress LLC Supplier No. HYW45144050MG
Butyne-DOTA-tris(t-butyl ester) is a bifunctional chelator and macrocyclic DOTA derivative used for bioconjugation and tumor pre-targeting research. It contains tert-butyl protected carboxylate arms and a terminal butyne handle that enables click-style conjugation to peptides, proteins, or targeting vectors, facilitating subsequent radiolabeling or imaging workflows. The material is supplied as a white to off-white solid with high reported purity and recommended cold storage for long-term stability.
  • Bifunctional chelator suitable for bioconjugation
  • Terminal alkyne handle enables click chemistry conjugation
  • Tert-butyl protected carboxylates improve stability during synthesis
  • High purity suitable for radiolabeling workflows
  • Available in multiple pack sizes for research needs
  • Recommended cold storage to preserve integrity