Learn More
Medchemexpress LLC Butyne-DOTA-tris(t-butyl ester) | 2125661-54-1 | 98.5% | 623.82 g·mol⁻¹ | C32H57N5O7 | 100 MG
SDP
Catalog No. 5000281723
Encompass_Preferred
Butyne-DOTA-tris(t-butyl ester) is a DOTA-derived bifunctional chelator featuring an alkyne functional group and tert-butyl protected carboxylates. It is intended for conjugation to peptides and radionuclides in radiopharmaceutical and imaging probe synthesis; the protected ester form simplifies handling and allows controlled deprotection to the metal-chelating DOTA core.
  • Bifunctional chelator for peptide and radionuclide conjugation.
  • Alkyne handle enables click chemistry.
  • Tert-butyl protected carboxylates allow controlled deprotection.
  • High HPLC purity (98.5%).
  • Available in research-scale quantities for probe synthesis.
Catalog No. 50-002-81723 Supplier Medchemexpress LLC Supplier No. HYW451440100MG
Butyne-DOTA-tris(t-butyl ester) is a DOTA-derived bifunctional chelator featuring an alkyne functional group and tert-butyl protected carboxylates. It is intended for conjugation to peptides and radionuclides in radiopharmaceutical and imaging probe synthesis; the protected ester form simplifies handling and allows controlled deprotection to the metal-chelating DOTA core.
  • Bifunctional chelator for peptide and radionuclide conjugation.
  • Alkyne handle enables click chemistry.
  • Tert-butyl protected carboxylates allow controlled deprotection.
  • High HPLC purity (98.5%).
  • Available in research-scale quantities for probe synthesis.