Learn More
Medchemexpress LLC Butyne-DOTA-tris(t-butyl ester) | 2125661-54-1 | 98.5% | 623.82 g/mol | C32H57N5O7 | 25 MG
SDP
Catalog No. 5000281108
Encompass_Preferred
Butyne-DOTA-tris(t-butyl ester) is a DOTA-based bifunctional chelator designed for conjugation of peptides and radionuclides. It is intended for radiopharmaceutical development and tumor pre-targeting workflows, supplied as a white to off-white solid.
  • Bifunctional DOTA derivative for peptide and radionuclide conjugation.
  • Supports tumor pre-targeting and radiopharmaceutical research.
  • High purity suitable for analytical and synthetic applications (98.5%).
  • Molecular weight 623.82 g/mol; formula C32H57N5O7.
  • Available in small research pack sizes, including 25 MG.
  • Provided with SDS and documentation for safe handling.
Catalog No. 50-002-81108 Supplier Medchemexpress LLC Supplier No. HYW45144025MG
Butyne-DOTA-tris(t-butyl ester) is a DOTA-based bifunctional chelator designed for conjugation of peptides and radionuclides. It is intended for radiopharmaceutical development and tumor pre-targeting workflows, supplied as a white to off-white solid.
  • Bifunctional DOTA derivative for peptide and radionuclide conjugation.
  • Supports tumor pre-targeting and radiopharmaceutical research.
  • High purity suitable for analytical and synthetic applications (98.5%).
  • Molecular weight 623.82 g/mol; formula C32H57N5O7.
  • Available in small research pack sizes, including 25 MG.
  • Provided with SDS and documentation for safe handling.