Learn More
Medchemexpress LLC Butyraldehyde 2,4-dinitrophenylhydrazone-d3 | 259824-54-9 | 99.0% | 255.25 | 1 MG
SDP
Catalog No. 5000451674
Encompass_Preferred
Butyraldehyde 2,4-dinitrophenylhydrazone-d3 is the deuterium labeled Butyraldehyde 2,4-dinitrophenylhydrazone. It is intended for research use only. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-51674 Supplier Medchemexpress LLC Supplier No. HYW009488S1MG
Butyraldehyde 2,4-dinitrophenylhydrazone-d3 is the deuterium labeled Butyraldehyde 2,4-dinitrophenylhydrazone. It is intended for research use only. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.