Learn More
Medchemexpress LLC Butyraldehyde 2,4-dinitrophenylhydrazone-d3 | 259824-54-9 | MFCD00223775 | 99.0% | 255.25 | 5 MG
SDP
Catalog No. 5000452053
Encompass_Preferred
Butyraldehyde 2,4-dinitrophenylhydrazone-d3 is the deuterium labeled Butyraldehyde 2,4-dinitrophenylhydrazone. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-52053 Supplier Medchemexpress LLC Supplier No. HYW009488S5MG
Butyraldehyde 2,4-dinitrophenylhydrazone-d3 is the deuterium labeled Butyraldehyde 2,4-dinitrophenylhydrazone. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.