Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butyraldehyde

Catalog No. 418102800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-810-2800ML 800 mL
41-810-2100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41-810-2800ML Supplier Sigma Aldrich Supplier No. 418102800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Butyraldehyde

Specifications

CAS 123-72-8
Melting Point -96°C (lit.)
Density 0.8 g/mL (at 25°C (literature))
Boiling Point 75°C (lit.)
Molecular Formula C4H8O
Refractive Index n20/D 1.380 (literature)
Linear Formula CH3CH2CH2CHO
MDL Number MFCD00007023
Quantity 800 mL
Synonym Butanal
Molecular Weight (g/mol) 72.11
Percent Purity ≥99.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.