Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyraldehyde, 99%

Catalog No. AC108091000
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108091000 100 mL Glass bottle
AC108090010 1 L Glass bottle
AC108090025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC108091000 Supplier Thermo Scientific Chemicals Supplier No. 108091000
Add to Cart
This item is not returnable. View return policy

CAS: 123-72-8 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-72-8
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00007023
InChI Key ZTQSAGDEMFDKMZ-UHFFFAOYSA-N
Synonym butyraldehyde, n-butyraldehyde, butyral, 1-butanal, butyric aldehyde, butanaldehyde, butaldehyde, butalyde, butal, n-butanal
PubChem CID 261
ChEBI CHEBI:15743
IUPAC Name butanal
SMILES CCCC=O

Specifications

Melting Point -96.0°C
Color Colorless
Density 0.8170g/mL
Boiling Point 75.0°C
Flash Point −12°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3770 to 1.3830
Linear Formula CH3CH2CH2CHO
Quantity 100 mL
Beilstein 01, 662
Specific Gravity 0.817
Merck Index 15, 1595
Solubility Information Solubility in water: 7.1% (25°C). Other solubilities: miscible with ethanol, ether, ethyl acetate,, acetone and toluene
Viscosity 0.43 mPa.s (20°C)
Formula Weight 72.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Butyraldehyde
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber 204-646-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.