Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Butyramide | 541-35-5 | MFCD00041894 | 98.1% | 87.12 g/mol | C4H9NO | 50 G
SDP

Catalog No. 5000208454
Encompass_Preferred

Butyramide (C4H9NO, MW 87.12 g/mol) is a biochemical reagent intended for research use as an organic compound or biological material.

  • Cas number: 541-35-5.
  • Purity: 98.1% (rounded from detailed spec).
  • Molecular weight: 87.12 g/mol.
  • Chemical formula: C4H9NO.
  • Available pack sizes: 25 G, 50 G, 100 G, 500 G.
  • Intended use: research use only.

Catalog No. 50-002-08454 Supplier Medchemexpress LLC Supplier No. HYW08831950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Butyramide (C4H9NO, MW 87.12 g/mol) is a biochemical reagent intended for research use as an organic compound or biological material.

  • Cas number: 541-35-5.
  • Purity: 98.1% (rounded from detailed spec).
  • Molecular weight: 87.12 g/mol.
  • Chemical formula: C4H9NO.
  • Available pack sizes: 25 G, 50 G, 100 G, 500 G.
  • Intended use: research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.