Learn More
Key Scientific Products Butyrate Discs, Bulk Pack, 50 Tests per Pack
SDP
Catalog No. NC9803274
Encompass_Preferred
Differentiate Branhamella catarrhalis from Neisseria spp. by demonstrating the enzymatic hydrolysis of bromo-chloro-indolyl butyrate. Includes 50 tests per pack.
Catalog No. NC9803274 Supplier Key Scientific Products Supplier No. K9131B
Differentiate Branhamella catarrhalis from Neisseria spp. by demonstrating the enzymatic hydrolysis of bromo-chloro-indolyl butyrate. Includes 50 tests per pack.