Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyric acid, 99+%

Catalog No. aal13189ae
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13189AE 100 mL
AAL13189AP 500 mL
AAL131890F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL13189AE Supplier Thermo Scientific Chemicals Supplier No. L13189AE
Only null left
Add to Cart

CAS: 107-92-6 | C4H8O2 | 88.11 g/mol

Butyric acid is a useful as precursor for the preparation of various butyrate esters. It is used as a food and perfume additive. It is also used as an animal feed supplement. Due to its powerful odor, it has also been used as a fishing bait additive and as a stink bomb. It is widely used as a flavoring agent for human food and beverages.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyric acid is a useful as precursor for the preparation of various butyrate esters. It is used as a food and perfume additive. It is also used as an animal feed supplement. Due to its powerful odor, it has also been used as a fishing bait additive and as a stink bomb. It is widely used as a flavoring agent for human food and beverages.

Solubility
Miscible with water, ethanol and ether. Slightly miscible with carbon tetrachloride.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-92-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002814
InChI Key FERIUCNNQQJTOY-UHFFFAOYSA-N
Synonym butyric acid, n-butyric acid, n-butanoic acid, propylformic acid, ethylacetic acid, butanic acid, 1-propanecarboxylic acid, butyrate, 1-butyric acid, buttersaeure
PubChem CID 264
ChEBI CHEBI:30772
IUPAC Name butanoic acid
SMILES CCCC(O)=O

Specifications

pH 2.5
Melting Point -8°C
Boiling Point 162°C to 164°C
Linear Formula CH3CH2CH2COOH
UN Number UN2820
Beilstein 906770
Merck Index 14,1593
Solubility Information Miscible with water,ethanol and ether. Slightly miscible with carbon tetrachloride.
Formula Weight 88.11
Percent Purity ≥99%
Density 0.964
Quantity 100 mL
Flash Point 69°C (156°F)
Odor Rancid
Refractive Index 1.398
Chemical Name or Material Butyric acid
Show More Show Less
Hazard Category H227-H302-H314-H335
Hazard Statement GHS H Statement
H311-H314-H318-H227
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: BUTYRIC ACID
EINECSNumber 203-532-3
RTECSNumber ES5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.