Learn More
Cayman Chemical Butyric Acid-d7, Purity: ≥98% deuterated forms (d1-d7), CAS: 73607-83-7, 100 mg
SDP
Catalog No. 502989711
Encompass_Preferred
An internal standard for the quantification of sodium butyrate by GC- or LC-MS
Catalog No. 50-298-9711 Supplier Cayman Chemical Supplier No. 29408100 MG
An internal standard for the quantification of sodium butyrate by GC- or LC-MS