Learn More
Cayman Chemical Butyric Acid-d7, Purity: ≥98% deuterated forms (d1-d7), CAS: 73607-83-7, 500 mg
SDP
Catalog No. 502988764
Encompass_Preferred
An internal standard for the quantification of sodium butyrate by GC- or LC-MS
Catalog No. 50-298-8764 Supplier Cayman Chemical Supplier No. 29408500 MG
An internal standard for the quantification of sodium butyrate by GC- or LC-MS