Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyric acid sodium salt, 98%

Catalog No. ac263190050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC263190050 5 g Glass bottle
AC263190250 25 g Glass bottle
AC263191000 100 g Glass bottle
AC263195000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC263190050 Supplier Thermo Scientific Chemicals Supplier No. 263190050
Only null left
Add to Cart

CAS: 156-54-7 | C4H7NaO2 | 110.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-54-7
Molecular Formula C4H7NaO2
Molecular Weight (g/mol) 110.09
MDL Number MFCD00002816
InChI Key MFBOGIVSZKQAPD-UHFFFAOYSA-M
Synonym sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1
PubChem CID 5222465
ChEBI CHEBI:64103
SMILES [Na+].CCCC([O-])=O

Specifications

Melting Point 250.0°C to 253.0°C
Color White
Assay Percent Range 98%
Linear Formula CH3CH2CH2COONa
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in ether
IUPAC Name sodium;butanoate
Formula Weight 110.09
Percent Purity 99%
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Amorphous Powder
Chemical Name or Material Butyric acid sodium salt
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.