Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butyric anhydride

Catalog No. B103551 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B103551 500 mL
B1035511L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B103551 Supplier Sigma Aldrich Supplier No. B103551500ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Butyric anhydride

Specifications

Melting Point -75°C to +66°C (lit.)
CAS 106-31-0
Boiling Point 198°C to 199°C (lit.)
Molecular Formula C8H14O3
Linear Formula (CH3CH2CH2CO)2 O
MDL Number MFCD00009389
Synonym Butanoic anhydride
Molecular Weight (g/mol) 158.19
Percent Purity 98%
Density 0.967 g/mL (at 25°C (literature))
Quantity 500 mL
Refractive Index n20/D 1.413 (literature)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.