Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyroin 96.0+%, TCI America™
SDP

Catalog No. B097125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B097125ML 25 mL
B09715ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B097125ML Supplier TCI America Supplier No. B097125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 496-77-5
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00021928
InChI Key BVEYJWQCMOVMAR-UHFFFAOYNA-N
Synonym butyroin, 5-hydroxy-4-octanone, 4-octanone, 5-hydroxy, 5-octanol-4-one, octan-4-ol-5-one, fema no. 2587, 5-hydroxy-octan-4-one, acmc-1aijr, ksc236m9n, +/--5-hydroxy-4-octanone
PubChem CID 219794
IUPAC Name 5-hydroxyoctan-4-one
SMILES CCCC(O)C(=O)CCC

Specifications

Color Yellow
Boiling Point 96°C
Quantity 25 mL
Formula Weight 144.21
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyroin
EINECSNumber (2)-0595
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.